missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | CEL |
|---|---|
| Concentration | 0.23 mg/mL |
| Applications | Immunofluorescence, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16814544
|
Proteintech
15384-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16804544
|
Proteintech
15384-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CEL catalyzes fat and vitamin absorption. CEL acts in concert with pancreatic lipase and colipase for the complete digestion of dietary triglycerides.The protein encoded by this gene is a glycoprotein secreted from the pancreas into the digestive tract and from the lactating mammary gland into human milk. The physiological role of this protein is in cholesterol and lipid-soluble vitamin ester hydrolysis and absorption. This encoded protein promotes large chylomicron production in the intestine. Also its presence in plasma suggests its interactions with cholesterol and oxidized lipoproteins to modulate the progression of atherosclerosis. In pancreatic tumoral cells, this encoded protein is thought to be sequestrated within the Golgi compartment and is probably not secreted. This gene contains a variable number of tandem repeat (VNTR) polymorphism in the coding region that may influence the function of the encoded protein.Specifications
| CEL | |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| P07882, P19835, Q64285 | |
| 1056, 12613, 24254 | |
| CEL Fusion Protein Ag5262 | |
| Primary | |
| -20°C | |
| CEL |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| BAL, Bile salt stimulated lipase, Bile salt-activated lipase, BSDL, BSSL, Bucelipase, Carboxyl ester lipase, CEase, CEL, CELL, Cholesterol esterase, FAP, FAPP, LIPA, MODY8, Pancreatic lysophospholipase, Sterol esterase | |
| CEL | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title