Learn More
CEL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15384-1-AP-20UL
 View more versions of this product
Product Code. 16814544
  • 145.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
CEL catalyzes fat and vitamin absorption. CEL acts in concert with pancreatic lipase and colipase for the complete digestion of dietary triglycerides.The protein encoded by this gene is a glycoprotein secreted from the pancreas into the digestive tract and from the lactating mammary gland into human milk. The physiological role of this protein is in cholesterol and lipid-soluble vitamin ester hydrolysis and absorption. This encoded protein promotes large chylomicron production in the intestine. Also its presence in plasma suggests its interactions with cholesterol and oxidized lipoproteins to modulate the progression of atherosclerosis. In pancreatic tumoral cells, this encoded protein is thought to be sequestrated within the Golgi compartment and is probably not secreted. This gene contains a variable number of tandem repeat (VNTR) polymorphism in the coding region that may influence the function of the encoded protein.
Specifications
Specifications
CEL
Polyclonal
Unconjugated
CEL
BAL, Bile salt stimulated lipase, Bile salt-activated lipase, BSDL, BSSL, Bucelipase, Carboxyl ester lipase, CEase, CEL, CELL, Cholesterol esterase, FAP, FAPP, LIPA, MODY8, Pancreatic lysophospholipase, Sterol esterase
Rabbit
Antigen Affinity Chromatography
RUO
1056, 12613, 24254
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07882, P19835, Q64285
CEL
CEL Fusion Protein Ag5262
20 μL
Primary
Rat, Mouse, Human