missing translation for 'onlineSavingsMsg'
Learn More

CDKI-73, MedChemExpress

Product Code. p-200098312
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485168

Brand: Medchem Express HY12445

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CDKI-73 (LS-007) is an orally active and highly efficacious CDK9 inhibitor, with Ki values of 4 nM, 4 nM and 3 nM for CDK9, CDK1 and CDK2, respectively. CDKI-73 down-regulates the RNAPII phosphorylation. CDKI-73 is also a novel pharmacological inhibitor of Rab11 cargo delivery and innate immune secretion.

CDKI-73 (LS-007) is an orally active and highly efficacious CDK9 inhibitor, with Ki values of 4 nM, 4 nM and 3 nM for CDK9, CDK1 and CDK2, respectively. CDKI-73 down-regulates the RNAPII phosphorylation. CDKI-73 is also a novel pharmacological inhibitor of Rab11 cargo delivery and innate immune secretion.

Specifications

Chemical Name or Material CDKI-73
CAS 1421693-22-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H15FN6O2S2
Quantity 10 mg
Synonym LS-007
Solubility Information DMSO : ≥ 52 mg/mL (131.83 mM)
SMILES CNC1=NC(C)=C(C2=C(F)C=NC(NC3=CC(S(=O)(N)=O)=CC=C3)=N2)S1
Molecular Weight (g/mol) 394.45
Formula Weight 394.45
Percent Purity 98.36%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.