Learn More
Abnova™ CDK7/CEN5q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0181.200uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16043285
  • 2210.00 € / 200µL
Estimated Shipment: 27-10-2022
to see stock.
Add to basket
Includes: DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Description
Description
Specifications
Specifications
Liquid
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
FISH-Ce
Store at 4°C in the dark.
200 μL
1022
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe