missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CDK7/CEN5q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
1475.00 € - 2430.00 €
Specifications
Content And Storage | Store at 4°C in the dark. |
---|---|
Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
Format | Tube |
For Use With (Application) | FISH-Ce |
Gene ID (Entrez) | 1022 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16033285
|
Abnova™
FG0181.100UL |
100 μL |
1475.00 €
100µL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043285
|
Abnova™
FG0181.200UL |
200 μL |
2430.00 €
200µL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at 4°C in the dark. | |
Tube | |
1022 | |
Liquid |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
FISH-Ce | |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |