missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC25B Antibody (3D6), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00 € - 533.00 €
Specifications
| Antigen | CDC25B |
|---|---|
| Clone | 3D6 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043828
|
Novus Biologicals
NBP3-26672-50ul |
50 μL |
343.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043840
|
Novus Biologicals
NBP3-26672-100ul |
100 μL |
533.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDC25B Monoclonal antibody specifically detects CDC25B in Human samples. It is validated for ELISASpecifications
| CDC25B | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CDC25HU2, cell division cycle 25 homolog B (S. cerevisiae), cell division cycle 25 homolog B (S. pombe), cell division cycle 25B, Dual specificity phosphatase Cdc25B, EC 3.1.3.48, M-phase inducer phosphatase 2 | |
| A synthesized peptide derived from Human CDC25B [UniProt P30305] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 3D6 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Phospho Specific | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 994 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title