missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC25B Antibody (3D6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26672-50ul
This item is not returnable.
View return policy
Description
CDC25B Monoclonal antibody specifically detects CDC25B in Human samples. It is validated for ELISA
Specifications
| CDC25B | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 3D6 | |
| ELISA | |
| CDC25HU2, cell division cycle 25 homolog B (S. cerevisiae), cell division cycle 25 homolog B (S. pombe), cell division cycle 25B, Dual specificity phosphatase Cdc25B, EC 3.1.3.48, M-phase inducer phosphatase 2 | |
| A synthesized peptide derived from Human CDC25B [UniProt P30305] | |
| 50 μL | |
| Cancer, Cell Cycle and Replication, Phospho Specific | |
| 994 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction