missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDC123 Recombinant Rabbit Monoclonal Antibody (23GB3755), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
186.00 € - 446.00 €
Specifications
| Antigen | CDC123 |
|---|---|
| Clone | 23GB3755 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089311
![]() |
Invitrogen™
MA552630 |
100 μL |
446.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089651
|
Invitrogen™
MA552631 |
20 μL |
186.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDC123 Recombinant Monoclonal antibody specifically detects CDC123 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| CDC123 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O75794 | |
| 8872 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3755 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AA959893; C10orf7; Cdc123; cell division cycle 123; cell division cycle 123 homolog; cell division cycle protein 123 homolog; Cell division cycle protein 123-like protein; D123; D123 gene product; G431001I09Rik; HT-1080; hypothetical protein; Protein D123; PZ32; QccE-12339 | |
| A synthetic peptide of human CDC123 (11-70AA). | |
| Primary | |
| -20°C | |
| CDC123 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title