Learn More
Invitrogen™ CDC123 Recombinant Rabbit Monoclonal Antibody (23GB3755), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA552631
 View more versions of this product
Product Code. 30089651
  • 186.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
The BCKDHA gene encodes the E1-alpha subunit of the branched-chain alpha-keto acid. The BCKD complex consists of 3 catalytic components: a heterotetrameric (alpha2-beta2) branched-chain alpha-keto acid decarboxylase (E1), a homo-24-meric dihydrolipoyl transacylase, and a homodimeric dihydrolipoamide dehydrogenase. E1 is a thiamine pyrophosphate (TPP)-dependent enzyme. The reaction is irreversible and constitutes the first committed step in BCAA oxidation. The BCKDHB gene encodes the beta subunit of E1. The complex also contains 2 regulatory enzymes, a kinase and a phosphorylase.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CDC123
Recombinant Monoclonal
Unconjugated
CDC123
AA959893; C10orf7; Cdc123; cell division cycle 123; cell division cycle 123 homolog; cell division cycle protein 123 homolog; Cell division cycle protein 123-like protein; D123; D123 gene product; G431001I09Rik; HT-1080; hypothetical protein; Protein D123; PZ32; QccE-12339
Rabbit
Affinity chromatography
RUO
8872
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB3755
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O75794
CDC123
A synthetic peptide of human CDC123 (11-70AA).