Learn More
Paragon Scientific™ CCS Viscosity Standards
Product Code. 15588854
Change view
Click to view available options
Type:
SAE Specification J308
SAE Specification J309
SAE Specification J310
SAE Specification J311
SAE Specification J312
SAE Specification J314
SAE Specification J315
SAE Specification J316
SAE Specification J317
SAE Specification J318
SAE Specification J319
SAE Specification J320
SAE Specification J321
SAE Specification J322
SAE Specification J323
SAE Specification J324
SAE Specification J325
SAE Specification J326
SAE Specification J328
Unit Size:
500mL
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
15588854 SAE Specification J308 500mL
15598854 SAE Specification J309 500mL
15508864 SAE Specification J310 500mL
15518864 SAE Specification J311 500mL
15528864 SAE Specification J312 500mL
15538864 SAE Specification J314 500mL
15548864 SAE Specification J315 500mL
15558864 SAE Specification J316 500mL
15568864 SAE Specification J317 500mL
15578864 SAE Specification J318 500mL
15588864 SAE Specification J319 500mL
15598864 SAE Specification J320 500mL
15508874 SAE Specification J321 500mL
15518874 SAE Specification J322 500mL
15528874 SAE Specification J323 500mL
15538874 SAE Specification J324 500mL
15548874 SAE Specification J325 500mL
15558874 SAE Specification J326 500mL
15523556 SAE Specification J328 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Product Code. 15588854 Supplier Paragon Scientific™ Supplier No. CL140
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Paragon ISO 17025 / ISO 17034 certified CCS viscosity standards are for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) testing to ASTM D5293 and SAE Specification J300.
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Full Traceability to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Manufactured in the United Kingdom
  • 2 year shelf life
Specifications
Certifications/Compliance Full Traceability to National Standards
Type SAE Specification J308
Quantity 500 mL
Nominal values quoted