Learn More
Paragon Scientific™ CCS Viscosity Standards
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Full Traceability to National Standards
- Ensures full compliance to ASTM & IP test method protocol
- Manufactured in the United Kingdom
- 2 year shelf life
Specifications
Specifications
| Certifications/Compliance | Full Traceability to National Standards |
| Type | SAE Specification J308 |
| Quantity | 500 mL |
Nominal values quoted