Learn More
CCR4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-86584
This item is not returnable.
View return policy
Description
CCR4 Polyclonal specifically detects CCR4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| CCR4 | |
| RUO |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20-1:50 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LQIYSADTPSSSYTQSTMDHDLHDAL | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |