missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
375.00 € - 573.00 €
Specifications
| Antigen | CCR4 |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20-1:50 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18405451
|
Novus Biologicals
NBP1-86584-25ul |
25 μL |
375.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18776263
|
Novus Biologicals
NBP1-86584 |
573.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
CCR4 Polyclonal specifically detects CCR4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CCR4 | |
| Polyclonal | |
| Rabbit | |
| Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5 | |
| CCR4 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20-1:50 | |
| Unconjugated | |
| RUO | |
| Human | |
| P51679 | |
| 1233 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LQIYSADTPSSSYTQSTMDHDLHDAL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title