missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caveolin-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
200.00 €
Specifications
| Antigen | Caveolin-1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 - 2 μg/mL, Flow Cytometry 3 - 5 μg 106 cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 2 μg/mL, Immunoprecipitation 5 μg |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Caveolin-1 Polyclonal specifically detects Caveolin-1 in Human, Mouse, Rat, Canine, Hamster samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| Caveolin-1 | |
| Western Blot 1 - 2 μg/mL, Flow Cytometry 3 - 5 μg 106 cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 2 μg/mL, Immunoprecipitation 5 μg | |
| Polyclonal | |
| Rabbit | |
| Caveolae Markers, Cellular Markers, Lipid and Metabolism, Lipid Droplets | |
| PBS with 1 mg/ml BSA with 0.05% Sodium Azide | |
| CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085 | |
| CAV1 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Q03135 | |
| 857 | |
| Synthetic peptide corresponding to residues M(1) S G G K Y V D S E G H L Y T V P(17) C of human CAV1. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title