missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caveolin-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB300-509-0.025mg
This item is not returnable.
View return policy
Description
Caveolin-1 Polyclonal specifically detects Caveolin-1 in Human, Mouse, Rat, Canine, Hamster samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| Caveolin-1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 1 mg/ml BSA with 0.05% Sodium Azide | |
| CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 - 2 μg/mL, Flow Cytometry 3 - 5 μg 106 cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 2 μg/mL, Immunoprecipitation 5 μg | |
| Q03135 | |
| CAV1 | |
| Synthetic peptide corresponding to residues M(1) S G G K Y V D S E G H L Y T V P(17) C of human CAV1. | |
| 0.025 mg | |
| Caveolae Markers, Cellular Markers, Lipid and Metabolism, Lipid Droplets | |
| 857 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction