Learn More
Carboxyphosphamide Benzyl Ester, TRC
Product Code. 30367554
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367554 25 mg -
30358470 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30367554 Supplier Toronto Research Chemicals Supplier No. TRCC18135525MG
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 37979-67-2
Molecular Formula C14H21Cl2N2O4P
Molecular Weight (g/mol) 383.21
Synonym 3-[[Amino[bis(2-chloroethyl)amino]phosphinyl]oxy]propanoic Acid Phenylmethyl Ester, 3-[[Amino[bis(2-chloroethyl)amino]phosphinyl]oxy]propanoic Acid Benzyl Ester
IUPAC Name benzyl 3-[amino-[bis(2-chloroethyl)amino]phosphoryl]oxypropanoate
SMILES NP(=O)(OCCC(=O)OCc1ccccc1)N(CCCl)CCCl
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C14H21Cl2N2O4P/c15-7-9-18(10-8-16)23(17,20)22-11-6-14(19)21-12-13-4-2-1-3-5-13/h1-5H,6-12H2,(H2,17,20)
Formula Weight 382.0616
Chemical Name or Material Carboxyphosphamide Benzyl Ester