CAS RN 37979-67-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Carboxyphosphamide Benzyl Ester, TRC
CAS: 37979-67-2 Molecular Formula: C14H21Cl2N2O4P Molecular Weight (g/mol): 383.21 Synonym: 3-[[Amino[bis(2-chloroethyl)amino]phosphinyl]oxy]propanoic Acid Phenylmethyl Ester,3-[[Amino[bis(2-chloroethyl)amino]phosphinyl]oxy]propanoic Acid Benzyl Ester IUPAC Name: benzyl 3-[amino-[bis(2-chloroethyl)amino]phosphoryl]oxypropanoate SMILES: NP(=O)(OCCC(=O)OCc1ccccc1)N(CCCl)CCCl