missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calibration Buffer pH 1.68, Hanna Instruments™
Click to view available options
Quantity:
500 mL
Unit Size:
460mL
Specifications
Specifications
| Color | Colorless |
| pH | 1.68 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solution Type | Buffer |
| Formulation | >1% to <5% Potassium Tetraoxalate Dihydrate |
| Odor | Odorless |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction