Learn More
Calcitrol 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30310266
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30310266 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30310266 Supplier Dr. Ehrenstorfer Supplier No. DREA10934950AL100
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Calcitriol
CAS 32222-06-3
Analyte or Component Names Calcitriol
Shipping Condition Room Temperature
Molecular Formula C27 H44 O3
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Calcitriol, (5Z,7E)-9,10-Secocholesta-5,7,10(19)-triene-1alpha,3beta,25-triol
InChI Formula InChI=1S/C27H44O3/c1-18(8-6-14-26(3,4)30)23-12-13-24-20(9-7-15-27(23,24)5)10-11-21-16-22(28)17-25(29)19(21)2/h10-11,18,22-25,28-30H,2,6-9,12-17H2,1,3-5H3/b20-10+,21-11-/t18-,22-,23-,24+,25+,27-/m1/s1
SMILES C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)C[C@H](O)C3=C)\CCC[C@]12C
IUPAC Name (1R,3S,5Z)-5-[(2E)-2-[(1R,3aS,7aR)-1-[(2R)-6-hydroxy-6-methylheptan-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexane-1,3-diol
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 416.64
Quantity 1 mL
Formula Weight 416.329 g/mol