missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cadherin-10 Antibody (SR4178), Novus Biologicals™
Rabbit Monoclonal Antibody
316.00 € - 382.00 €
Specifications
| Antigen | Cadherin-10 |
|---|---|
| Clone | SR4178 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041782
|
Novus Biologicals™
NBP3-27821-50ul |
50 μL |
316.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041663
|
Novus Biologicals™
NBP3-27821-100ul |
100 μL |
382.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cadherin-10 Monoclonal antibody specifically detects Cadherin-10 in Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
| Cadherin-10 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| cadherin 10, type 2 (T2-cadherin), T2-cadherin | |
| A synthesized peptide derived from human Cadherin-10 (Uniprot #: Q9Y6N8) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR4178 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 1008 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title