missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cadherin-10 Antibody (SR4178), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27821-100ul
This item is not returnable.
View return policy
Description
Cadherin-10 Monoclonal antibody specifically detects Cadherin-10 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| Cadherin-10 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR4178 | |
| Western Blot 1:500-1:2000 | |
| cadherin 10, type 2 (T2-cadherin), T2-cadherin | |
| A synthesized peptide derived from human Cadherin-10 (Uniprot #: Q9Y6N8) | |
| 100 μL | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| 1008 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction