missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2orf51 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | C2orf51 |
|---|---|
| Concentration | 0.24 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16824045
|
Proteintech
25464-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16814045
|
Proteintech
25464-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C2orf51, also named as TEX37 and TSC21, is a protein with calculated MW 21 kDa. We always detect 35 kDa in our WB detection.Specifications
| C2orf51 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q96LM6, Q9DAG4 | |
| 200523, 74221 | |
| C2orf51 Fusion Protein Ag21923 | |
| Primary | |
| -20°C | |
| TEX37 |
| 0.24 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| C2orf51, Protein TSC21, TEX37, TSC21 | |
| Tex37 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title