Learn More
C2orf51 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25464-1-AP-150UL
This item is not returnable.
View return policy
Description
C2orf51, also named as TEX37 and TSC21, is a protein with calculated MW 21 kDa. We always detect 35 kDa in our WB detection.Specifications
| C2orf51 | |
| Polyclonal | |
| Unconjugated | |
| TEX37 | |
| C2orf51, Protein TSC21, TEX37, TSC21 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 200523, 74221 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96LM6, Q9DAG4 | |
| Tex37 | |
| C2orf51 Fusion Protein Ag21923 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |