missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Abl Antibody (8E9), Novus Biologicals™
Mouse Monoclonal Antibody
218.00 € - 714.00 €
Specifications
| Antigen | c-Abl |
|---|---|
| Clone | 8E9 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:100, Immunoprecipitation 2 μg/mg, Immunohistochemistry-Paraffin 2 - 4 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18670729
|
Novus Biologicals
NBP2-49695-100ul |
100 μg |
218.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698816
|
Novus Biologicals
NBP2-49695 |
500 μg |
714.00 €
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
c-Abl Monoclonal antibody specifically detects c-Abl in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| c-Abl | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, DNA Repair, Signal Transduction, Tumor Suppressors | |
| PBS (pH 7.4), 0.2% BSA | |
| 25 | |
| IgG1 | |
| Protein G purified |
| 8E9 | |
| Western Blot 1 μg/mL, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:100, Immunoprecipitation 2 μg/mg, Immunohistochemistry-Paraffin 2 - 4 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Abelson murine leukemia viral oncogene homolog 1, ABL, c-ABL, c-abl oncogene 1, non-receptor tyrosine kinase, c-abl oncogene 1, receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.2, JTK7bcr/abl, p150bcr/c-abl oncogene protein, Proto-oncogene c-Abl, proto-oncogene tyrosine-protein kinase ABL1, tyrosine-protein kinase ABL1, v-abl, v-abl Abelson murine leukemia viral oncogene homolog 1 | |
| Recombinant Abl protein | |
| Primary | |
| Store at 4°C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title