missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Abl Antibody (8E9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-49695-100ul
This item is not returnable.
View return policy
Description
c-Abl Monoclonal antibody specifically detects c-Abl in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| c-Abl | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 μg/mL, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:100, Immunoprecipitation 2 μg/mg, Immunohistochemistry-Paraffin 2 - 4 μg/mL | |
| Abelson murine leukemia viral oncogene homolog 1, ABL, c-ABL, c-abl oncogene 1, non-receptor tyrosine kinase, c-abl oncogene 1, receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.2, JTK7bcr/abl, p150bcr/c-abl oncogene protein, Proto-oncogene c-Abl, proto-oncogene tyrosine-protein kinase ABL1, tyrosine-protein kinase ABL1, v-abl, v-abl Abelson murine leukemia viral oncogene homolog 1 | |
| Recombinant Abl protein | |
| 100 μg | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, DNA Repair, Signal Transduction, Tumor Suppressors | |
| 25 | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 8E9 | |
| Unconjugated | |
| PBS (pH 7.4), 0.2% BSA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur