Learn More
Butyl 3-(Butylamino)-4-phenoxy-5-sulfamoylbenzoate, Mikromol™
Description
Butyl 3-(Butylamino)-4-phenoxy-5-sulfamoylbenzoate
Specifications
Specifications
| Chemical Name or Material | Butyl 3-(Butylamino)-4-phenoxy-5-sulfamoylbenzoate |
| CAS | 32643-00-8 |
| Analyte or Component Names | Butyl 3-(Butylamino)-4-phenoxy-5-sulfamoylbenzoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H28 N2 O5 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Butyl 3-(Butylamino)-4-phenoxy-5-sulphamoylbenzoate, Bumetanide Imp. C (EP), Benzoic acid, 3-(aminosulfonyl)-5-(butylamino)-4-phenoxy-, butyl ester, Benzoic acid, 3-(butylamino)-4-phenoxy-5-sulfamoyl-, butyl ester (8CI), PF 1760, Bumetanide Impurity C |
| InChI Formula | InChI=1S/C21H28N2O5S/c1-3-5-12-23-18-14-16(21(24)27-13-6-4-2)15-19(29(22,25)26)20(18)28-17-10-8-7-9-11-17/h7-11,14-15,23H,3-6,12-13H2,1-2H3,(H2,22,25,26) |
| SMILES | CCCCNc1cc(cc(c1Oc2ccccc2)S(=O)(=O)N)C(=O)OCCCC |
| IUPAC Name | butyl 3-(butylamino)-4-phenoxy-5-sulfamoylbenzoate |
| Show More |