Learn More
Butyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate, Mikromol™
Description
Butyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate
Specifications
Specifications
| Chemical Name or Material | Butyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate |
| CAS | 2732346-94-8 |
| Analyte or Component Names | Butyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H28 Cl N O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Bezafibrate Imp. E (EP), Butyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate, Bezafibrate Impurity E |
| InChI Formula | InChI=1S/C20H28N2.H3O4P/c1-16(2)22(17(3)4)15-13-19(18-10-6-5-7-11-18)20-12-8-9-14-21-20;1-5(2,3)4/h5-12,14,16-17,19H,13,15H2,1-4H3;(H3,1,2,3,4) |
| SMILES | CCCCOC(=O)C(C)(C)Oc1ccc(CCNC(=O)c2ccc(Cl)cc2)cc1 |
| IUPAC Name | butyl 2-[4-[2-[(4-chlorobenzoyl)amino]ethyl]phenoxy]-2-methylpropanoate |
| Show More |