Learn More
Bunolol Hydrochloride, Mikromol™
Description
Bunolol Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Bunolol Hydrochloride |
| CAS | 31969-05-8 |
| Analyte or Component Names | Bunolol Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H25 N O3 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | 1(2H)-Naphthalenone, 5-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride (1:1), 1(2H)-Naphthalenone, 5-[3-[(1,1-dimethylethyl), 1(2H)-Naphthalenone, 5-[3-(tert-butylamino)-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride, (+/-)- (8CI), 1(2H)-Naphthalenone, 5-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride (9CI), 1(2H)-Naphthalenone, 5-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride, (+/-)-, (+/-)-Bunolol hydrochloride, Bunolol hydrochloride, W 6412A, dl-Bunolol hydrochloride |
| InChI Formula | InChI=1S/C17H25NO3.ClH/c1-17(2,3)18-10-12(19)11-21-16-9-5-6-13-14(16)7-4-8-15(13)20;/h5-6,9,12,18-19H,4,7-8,10-11H2,1-3H3;1H |
| SMILES | Cl.CC(C)(C)NCC(O)COc1cccc2C(=O)CCCc12 |
| IUPAC Name | 5-[3-(tert-butylamino)-2-hydroxypropoxy]-3,4-dihydro-2H-naphthalen-1-one;hydrochloride |
| Show More |