missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 4.01, METTLER TOLEDO™

Product Code. 10443131
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
1500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10443131 1000 mL 1000mL
10711765 250 mL 1500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10443131 Supplier Mettler Toledo Supplier No. 51340228
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The DKD (Deutscher Kalibrierdienst, German Calibration Service) certified buffers are the perfect product for regulated industries.

TRUSTED_SUSTAINABILITY

Specifications

Color Light Red
pH 4.01
Physical Form Liquid
Quantity 1000 mL
Solution Type pH Buffer
Odor None
Packaging 1000mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.