Learn More
Buffer solution, Citric Acid/Sodium Hydroxyde/Sodium Chloride, pH 4500mL, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
| Chemical Name or Material | Coloured Citric acid/ Sodium Hydroxide/ Sodium Chloride |
| pH | 4 |
| Quantity | 500 mL |
| Packaging | Bottle |
Safety and Handling
Product Identifier
- Buffer solution - pH 4.00