Learn More
Budesonide EP Impurity H, Mikromol™
Description
Budesonide EP Impurity H
Specifications
Specifications
| Chemical Name or Material | 16 alpha,17-[(1RS)-Butylidenebis(oxy)]-21-hydroxypregna-1,4,9(11)-triene-3,20-dione |
| CAS | 313474-58-7 |
| Analyte or Component Names | 16 alpha,17-[(1RS)-Butylidenebis(oxy)]-21-hydroxypregna-1,4,9(11)-triene-3,20-dione |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25H32O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 16α,17-[(1RS)-Butylidenebis(oxy)]-21-hydroxypregna-1,4,9(11)-triene-3,20-dione, Budesonide Imp. H (EP), 16alpha,17-[(1RS)-Butylidene-bis(oxy)]-21-hydroxypregna-1,4-diene-3,20-dione, (16alpha)-16,17-[Butylidenebis(oxy)]-21-hydroxy-pregna-1,4,9(11)-triene-3,20-dione, 9,11-Anhydrobudesonide, 16alpha,17-[(1RS)-Butylidenebis(oxy)]-21-hydroxypregna-1,4,9(11)-triene-3,20-dione, Budesonide Impurity H |
| SMILES | CCCC1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)C4=CC[C@]3(C)[C@@]2(O1)C(=O)CO |
| Molecular Weight (g/mol) | 412.52 |
| Quantity | 25 mg |