Learn More
Bromazine Hydrochloride, Mikromol™
Description
Bromazine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Bromazine Hydrochloride |
| CAS | 1808-12-4 |
| Analyte or Component Names | Bromazine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H20 Br N O . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Dimenhydrinate Imp. H (EP), Diphenhydramine Hydrochloride Imp. C (EP), Diphenhydramine Imp. C (EP), 2-[(RS)-(4-Bromophenyl)phenylmethoxy]-N,N-dimethylethanamine Hydrochloride, 4-Bromodiphenhydramine Hydrochloride, Bromazine Hydrochloride, Dimenhydrinate Imp. H (EP) as Hydrochloride, Diphenhydramine Hydrochloride Imp. C (EP) as Hydrochloride, Dimenhydrinate Impurity H as Hydrochloride, Diphenhydramine Hydrochloride Impurity C as Hydrochloride, Diphenhydramine Impurity C as Hydrochloride |
| InChI Formula | InChI=1S/C17H20BrNO.ClH/c1-19(2)12-13-20-17(14-6-4-3-5-7-14)15-8-10-16(18)11-9-15;/h3-11,17H,12-13H2,1-2H3;1H |
| SMILES | Cl.CN(C)CCOC(c1ccccc1)c2ccc(Br)cc2 |
| IUPAC Name | 2-[(4-bromophenyl)-phenylmethoxy]-N,N-dimethylethanamine;hydrochloride |
| Show More |