Learn More
Bromazine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Product Code. 30463490
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463490 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30463490 Supplier LoGiCal Supplier No. LGCAMP0640.0001
Please to purchase this item. Need a web account? Register with us today!
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bromazine Hydrochloride
CAS 1808-12-4
Analyte or Component Names Bromazine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C17 H20 Br N O . Cl H
Certifications/Compliance ISO 17034
Synonym Dimenhydrinate Imp. H (EP), Diphenhydramine Hydrochloride Imp. C (EP), Diphenhydramine Imp. C (EP), 2-[(RS)-(4-Bromophenyl)phenylmethoxy]-N,N-dimethylethanamine Hydrochloride, 4-Bromodiphenhydramine Hydrochloride, Bromazine Hydrochloride, Dimenhydrinate Imp. H (EP) as Hydrochloride, Diphenhydramine Hydrochloride Imp. C (EP) as Hydrochloride, Dimenhydrinate Impurity H as Hydrochloride, Diphenhydramine Hydrochloride Impurity C as Hydrochloride, Diphenhydramine Impurity C as Hydrochloride
InChI Formula InChI=1S/C17H20BrNO.ClH/c1-19(2)12-13-20-17(14-6-4-3-5-7-14)15-8-10-16(18)11-9-15;/h3-11,17H,12-13H2,1-2H3;1H
Solution Type Single Solution
SMILES Cl.CN(C)CCOC(c1ccccc1)c2ccc(Br)cc2
IUPAC Name 2-[(4-bromophenyl)-phenylmethoxy]-N,N-dimethylethanamine;hydrochloride
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 370.71
Quantity 1 mL
Formula Weight 369.0495 g/mol