missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRCA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP1-88361-25ul
This item is not returnable.
View return policy
Description
BRCA2 Polyclonal specifically detects BRCA2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| BRCA2 | |
| Polyclonal | |
| Western Blot Reported in scientific literature (PMID: 30250272)., Flow Cytometry Reported in scientific literature (PMID: 28904740)., Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin Reported in scientific literature ( PMID: 32529018). | |
| P51587 | |
| BRCA2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VHPISLSSSKCHDSVVSMFKIENHNDKTVSEKNNKCQLILQNNIEMTTGTFVEEITENYKRNTENEDNKYTAASRNSHNLEFDGSDSSKNDTVCIHKDETDLLFTDQHNICLKLSGQFMKEGNTQIKEDLS | |
| 25 μL | |
| Breast Cancer, Cancer | |
| 675 | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| BRCA1/BRCA2-containing complex, subunit 2, BRCC2, breast and ovarian cancer susceptibility gene, early onset, breast cancer 2 tumor suppressor, breast cancer 2, early onset, breast cancer susceptibility protein BRCA2, breast cancer type 2 susceptibility protein, BROVCA2, FACD, FAD, FAD1, FANCB, FANCD, FANCD1Fanconi anemia, complementation group D1, Fanconi anemia group D1 protein, GLM3, PNCA2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction