Learn More
BrandTech™ Transferpette™ S Pipet Accessories
Two replacement tip cones for BRAND Transferpette electronic and Transferpette S multichannel pipettes with 2 O-ring design
Brand:  BrandTech™ 705683
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10002452
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Instruments with a single O-ring must replace all tip cones
Specifications
Specifications
Transferpette S Pipet
Seal, V-ring and mounting tool