Learn More
BrandTech™ Transferpette™ S Pipet Accessories
Two replacement tip cones for BRAND Transferpette electronic and Transferpette S multichannel pipettes with 2 O-ring design
16.20 € - 17.35 €
Products 2
Product Code Brand Price Quantity & Availability  
Product Code Brand Price Quantity & Availability  
10276042
View Documents Promotion Details
BrandTech™
705684
300μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10002452
View Documents Promotion Details
BrandTech™
705683
200μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Instruments with a single O-ring must replace all tip cones