Learn More
BrandTech™ Transferpette™ S Pipet Accessories
Two replacement tip cones for BRAND Transferpette electronic and Transferpette S multichannel pipettes with 2 O-ring design
16.20 € - 17.35 €
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
10276042
|
BrandTech™
705684 |
300μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10002452
|
BrandTech™
705683 |
200μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Instruments with a single O-ring must replace all tip cones