missing translation for 'onlineSavingsMsg'
Learn More

Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate, 97%

Product Code. 11313936
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11313936

Brand: Thermo Scientific Alfa Aesar L17046.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 72022-68-5 | C22H40N2S10Zn | 718.556 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72022-68-5
Molecular Formula C22H40N2S10Zn
Molecular Weight (g/mol) 718.556
MDL Number MFCD00623815
InChI Key TXRIVAQWJHZFOT-UHFFFAOYSA-J
Synonym bis tetraethylammonium bis 2-thioxo-1,3-dithiole-4,5-dithiolato zincate
PubChem CID 15762389
IUPAC Name zinc;2-sulfanylidene-1,3-dithiole-4,5-dithiolate;tetraethylazanium
SMILES CC[N+](CC)(CC)CC.CC[N+](CC)(CC)CC.C1(=C(SC(=S)S1)[S-])[S-].C1(=C(SC(=S)S1)[S-])[S-].[Zn+2]

Specifications

Melting Point 202°C to 206°C
Quantity 1 g
Beilstein 4900651
Sensitivity Light sensitive
Formula Weight 718.6
Percent Purity 97%
Chemical Name or Material Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.