accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 72022-68-5

CH3NH3CH3CH3CCH3NH3CH3CH3CSSSSSSSSSSZn

CAS RN 72022-68-5

IUPAC Name: zinc(2+) bis((5-sulfanidyl-2-sulfanylidene-2H-1,3-dithiol-4-yl)sulfanide) bis(tetraethylazanium)
Synonyms: zinc(2+) bis((5-sulfanidyl-2-sulfanylidene-1,3-dithiol-4-yl)sulfanide) bis(tetraethylammonium)
Molecular Weight (g/mol): 718.56
Molecular Formula: C22H40N2S10Zn
InChi Key: TXRIVAQWJHZFOT-UHFFFAOYSA-J
SMILES: [Zn++].[S-]C1=C([S-])SC(=S)S1.[S-]C1=C([S-])SC(=S)S1.CC[N+](CC)(CC)CC.CC[N+](CC)(CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results