Learn More
Bis[4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl] (Cyclopentane-1,1-diyl)diacetate Dioxalate, Mikromol™
Description
Bis[4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl] (Cyclopentane-1,1-diyl)diacetate Dioxalate
Specifications
Specifications
| Chemical Name or Material | Bis[4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl] (Cyclopentane-1,1-diyl)diacetate Dioxalate |
| CAS | 2724726-70-7 |
| Analyte or Component Names | Bis[4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl] (Cyclopentane-1,1-diyl)diacetate Dioxalate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C33 H50 N8 O4 . 2 C2 H2 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Buspirone Hydrochloride Imp. H (EP), Buspirone Imp. H (EP), Buspirone Hydrochloride Imp. H (EP) as Dioxalate, Bis[4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl] (Cyclopentane-1,1-diyl)diacetate Dioxalate, Buspirone Hydrochloride Impurity H as Dioxalate, Buspirone Impurity H as Dioxalate |
| InChI Formula | InChI=1S/C12H18N2O3S.ClH/c1-4-5-13-6-9(15)14-10-8(2)7-18-11(10)12(16)17-3;/h7,13H,4-6H2,1-3H3,(H,14,15);1H |
| SMILES | OC(=O)C(=O)O.OC(=O)C(=O)O.O=C(CC1(CC(=O)OCCCCN2CCN(CC2)c3ncccn3)CCCC1)OCCCCN4CCN(CC4)c5ncccn5 |
| IUPAC Name | oxalic acid;4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl 2-[1-[2-oxo-2-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butoxy]ethyl]cyclopentyl]acetate |
| Show More |