missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation BioResolve-RP-mAb Column

Product Code. 15965922
Encompass
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
Diameter (Metric):
2.1 mm
4.6 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15965922

Brand: Waters Corporation 186008955

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

BioResolve RP mAb Columns contain superficially-porous-silica particles having an average pore diameter of 450 Å.

  • The particles are functionalized with a proprietary polyphenyl bonding process that was specifically designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 150 mm
Particle Size 2.7 μm
Pore Size 450 Å
Quantity 1/Ea.
Type Column
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.