Learn More
Bilastine, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Bilastine
Specifications
Specifications
| Chemical Name or Material | Bilastine |
| CAS | 202189-78-4 |
| Analyte or Component Names | Bilastine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C28 H37 N3 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 4-[2-[4-[1-(2-Ethoxyethyl)-1H-benzimidazol-2-yl]-1-piperidinyl]ethyl]-alpha,alpha-dimethylbenzeneacetic acid, Bilastine |
| InChI Formula | InChI=1S/C28H37N3O3/c1-4-34-20-19-31-25-8-6-5-7-24(25)29-26(31)22-14-17-30(18-15-22)16-13-21-9-11-23(12-10-21)28(2,3)27(32)33/h5-12,22H,4,13-20H2,1-3H3,(H,32,33) |
| SMILES | CCOCCn1c(nc2ccccc12)C3CCN(CCc4ccc(cc4)C(C)(C)C(=O)O)CC3 |
| IUPAC Name | 2-[4-[2-[4-[1-(2-ethoxyethyl)benzimidazol-2-yl]piperidin-1-yl]ethyl]phenyl]-2-methylpropanoic acid |
| Show More |