Learn More
Bifenazate, TRC
Product Code. 30006438
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006438 10 g 10g
30008992 25 g 25g
17982575 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006438 Supplier Toronto Research Chemicals Supplier No. TRCB38302010G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 149877-41-8
Molecular Formula C17 H20 N2 O3
Molecular Weight (g/mol) 300.35
Synonym 1-Methylethyl 2-(4-methoxy[1,1'-biphenyl]-3-yl)-hydrazinecarboxylate, Acramite, Bifenazate, D 2341, Floramite, Isopropyl 3-(4-methoxy-3-biphenylyl)carbazate
IUPAC Name propan-2-yl N-(2-methoxy-5-phenylanilino)carbamate
SMILES COc1ccc(cc1NNC(=O)OC(C)C)c2ccccc2
Specifications
Analyte or Component Names Bifenazate
Shipping Condition Room Temperature
InChI Formula InChI=1S/C17H20N2O3/c1-12(2)22-17(20)19-18-15-11-14(9-10-16(15)21-3)13-7-5-4-6-8-13/h4-12,18H,1-3H3,(H,19,20)
Solution Type Neat
Industry Type Environmental Testing
Quantity 10 g
Formula Weight 300.1474
Percent Purity >95%
Chemical Name or Material Bifenazate
Grade HPLC