Learn More
Betamethasone, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Betamethasone
Specifications
Specifications
| Chemical Name or Material | Betamethasone |
| CAS | 378-44-9 |
| Analyte or Component Names | Betamethasone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H29 F O5 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Betamethasone Acetate Imp. A (EP), Betamethasone Valerate Imp. A (EP), Dexamethasone Imp. B (EP), Betamethasone, 9-Fluoro-11beta,17,21-trihydroxy-16beta-methylpregna-1,4-diene-3,20-dione, Betamethasone Dipropionate Imp. A (Pharmeuropa), CCI 922, Ph Eur Betamethasone Valerate Impurity A, Betamethasone Sodium Phosphate related, CS valerate, Betamethasone, Betamethasone Acetate Impurity A, Betamethasone Dipropionate Impurity A, Betamethasone Valerate Impurity A, Dexamethasone Impurity B |
| InChI Formula | InChI=1S/C22H29FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15-,16-,17-,19-,20-,21-,22-/m0/s1 |
| SMILES | C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO |
| IUPAC Name | (8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one |
| Show More |