Learn More
Betahistine EP Impurity C (as Trimesilate), Mikromol™
Description
Betahistine EP Impurity C (as Trimesilate)
Specifications
Specifications
| Chemical Name or Material | N-Methyl-2-(pyridin-2-yl)-N-[2-(pyridin-2-yl)ethyl]ethanamine Trimesilate |
| CAS | 2731369-07-4 |
| Analyte or Component Names | N-Methyl-2-(pyridin-2-yl)-N-[2-(pyridin-2-yl)ethyl]ethanamine Trimesilate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H19 N3 . 3 C H4 O3 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Betahistine Dihydrochloride Imp. C (EP), Betahistine Imp. C (EP), N-Methyl-2-(pyridin-2-yl)-N-[2-(pyridin-2-yl)ethyl]ethanamine Trimesilate, Betahistine Dihydrochloride Imp. C (EP) as Trimesilate, Betahistine Dihydrochloride Impurity C as Trimesilate, Betahistine Impurity C as Trimesilate |
| InChI Formula | InChI=1S/C15H19N3.3CH4O3S/c1-18(12-8-14-6-2-4-10-16-14)13-9-15-7-3-5-11-17-15;3*1-5(2,3)4/h2-7,10-11H,8-9,12-13H2,1H3;3*1H3,(H,2,3,4) |
| SMILES | CN(CCc1ccccn1)CCc2ccccn2.CS(=O)(=O)O.CS(=O)(=O)O.CS(=O)(=O)O |
| IUPAC Name | methanesulfonic acid;N-methyl-2-pyridin-2-yl-N-(2-pyridin-2-ylethyl)ethanamine |
| Show More |