Learn More
Betahistine EP Impurity C (as Trimesilate), Mikromol™
Product Code. 30049859
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049859 25 mg 25mg
30052291 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30049859 Supplier Mikromol Supplier No. MM0587.040025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Betahistine EP Impurity C (as Trimesilate)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Methyl-2-(pyridin-2-yl)-N-[2-(pyridin-2-yl)ethyl]ethanamine Trimesilate
CAS 2731369-07-4
Analyte or Component Names N-Methyl-2-(pyridin-2-yl)-N-[2-(pyridin-2-yl)ethyl]ethanamine Trimesilate
Shipping Condition Room Temperature
Molecular Formula C15 H19 N3 . 3 C H4 O3 S
Certifications/Compliance ISO 9001
Synonym Betahistine Dihydrochloride Imp. C (EP), Betahistine Imp. C (EP), N-Methyl-2-(pyridin-2-yl)-N-[2-(pyridin-2-yl)ethyl]ethanamine Trimesilate, Betahistine Dihydrochloride Imp. C (EP) as Trimesilate, Betahistine Dihydrochloride Impurity C as Trimesilate, Betahistine Impurity C as Trimesilate
InChI Formula InChI=1S/C15H19N3.3CH4O3S/c1-18(12-8-14-6-2-4-10-16-14)13-9-15-7-3-5-11-17-15;3*1-5(2,3)4/h2-7,10-11H,8-9,12-13H2,1H3;3*1H3,(H,2,3,4)
SMILES CN(CCc1ccccn1)CCc2ccccn2.CS(=O)(=O)O.CS(=O)(=O)O.CS(=O)(=O)O
IUPAC Name methanesulfonic acid;N-methyl-2-pyridin-2-yl-N-(2-pyridin-2-ylethyl)ethanamine
Molecular Weight (g/mol) 529.65
Quantity 25 mg
Formula Weight 529.1222 g/mol