missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriethylammonium dichloroiodate, 97%, Thermo Scientific™

Product Code. 15428367
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428367 1 g 1g
15438367 5 g 5g
15448367 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15428367 Supplier Thermo Scientific Alfa Aesar Supplier No. H51872.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140410-47-5
Molecular Formula C13H22Cl2INO2
Molecular Weight (g/mol) 422.13
MDL Number MFCD11973847
InChI Key UYCBHUVBPDJSKR-UHFFFAOYSA-M
Synonym benzyltriethylammonium dichloroiodate
PubChem CID 121234138
IUPAC Name benzyl(triethyl)azanium;dichloro-oxido-oxo-$l^{5}-iodane
SMILES [O-][I](Cl)(Cl)=O.CC[N+](CC)(CC)CC1=CC=CC=C1

Specifications

Melting Point 79°C to 81°C
Linear Formula (CH3CH2)3N(CH2C6H5)ICl2
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 390.13
Percent Purity 97%
Chemical Name or Material Benzyltriethylammonium dichloroiodate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.