CAS RN 140410-47-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
422.13
Molecular Formula:
C13H22Cl2INO2
InChi Key:
UYCBHUVBPDJSKR-UHFFFAOYSA-M
SMILES:
[O-][I](Cl)(Cl)=O.CC[N+](CC)(CC)CC1=CC=CC=C1
1
–
1
of
1
results
1
Benzyltriethylammonium dichloroiodate, 97%, Thermo Scientific™
CAS: 140410-47-5 Molecular Formula: C13H22Cl2INO2 Molecular Weight (g/mol): 422.13 MDL Number: MFCD11973847 InChI Key: UYCBHUVBPDJSKR-UHFFFAOYSA-M Synonym: benzyltriethylammonium dichloroiodate PubChem CID: 121234138 IUPAC Name: benzyl(triethyl)azanium;dichloro-oxido-oxo-$l^{5}-iodane SMILES: [O-][I](Cl)(Cl)=O.CC[N+](CC)(CC)CC1=CC=CC=C1