missing translation for 'onlineSavingsMsg'
Learn More

Benzyloxyacetyl chloride, 95%

Product Code. p-7046441
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11343476

Brand: Thermo Scientific Alfa Aesar L14337.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 19810-31-2 | C9H9ClO2 | 184.619 g/mol

Benzyloxyacetyl chloride is a reagent used to construct substituted 2-azetidinones for further elaboration into annulated β-lactams. It is a commonly employed reagent for asymmetric synthesis of β-lactams. It is also used in preparation of (S)-3-(methylamino)-3-((R)-pyrrolidin-3-yl)propanenitrile, key intermediate in the preparation of fluoroquinolone antibiotic for respiratory tract infections, non-racemic helicene and β-lactams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyloxyacetyl chloride is a reagent used to construct substituted 2-azetidinones for further elaboration into annulated β-lactams. It is a commonly employed reagent for asymmetric synthesis of β-lactams. It is also used in preparation of (S)-3-(methylamino)-3-((R)-pyrrolidin-3-yl)propanenitrile, key intermediate in the preparation of fluoroquinolone antibiotic for respiratory tract infections, non-racemic helicene and β-lactams.

Solubility
Reacts with water.

Notes
It is sensitive to moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19810-31-2
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00010768
InChI Key QISAUDWTBBNJIR-UHFFFAOYSA-N
Synonym benzyloxyacetyl chloride, 2-benzyloxy acetyl chloride, benzyloxy acetyl chloride, benzyloxyacetylchloride, benzyloxy-acetyl chloride, benzyloxyacetic acid chloride, acetyl chloride, phenylmethoxy, acetyl chloride, benzyloxy, alpha-benzyloxy acetyl chloride, bnoch2cocl
PubChem CID 177085
IUPAC Name 2-phenylmethoxyacetyl chloride
SMILES C1=CC=C(C=C1)COCC(=O)Cl

Specifications

Density 1.189
Boiling Point 84°C to 87°C (0.4 mmHg)
Flash Point >110°C (230°F)
Linear Formula C6H5CH2OCH2COCl
Refractive Index 1.523
Quantity 1 g
UN Number UN3265
Beilstein 1947363
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 184.62
Percent Purity 95%
Chemical Name or Material Benzyloxyacetyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.