accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClOO

CAS RN 19810-31-2

IUPAC Name: 2-(benzyloxy)acetyl chloride
Synonyms: 2-(benzyloxy)acetyl chloride
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: QISAUDWTBBNJIR-UHFFFAOYSA-N
SMILES: ClC(=O)COCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results