missing translation for 'onlineSavingsMsg'
Learn More

Benzyl (2-((((2,2-Dimethyl-1,3-dioxolan-4-yl)methoxy)(hydroxy)phosphoryl)oxy)ethyl)carbamate Sodium Salt, TRC

Product Code. 30345721
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
missing translation for 'unitSize'
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30345721 5 mg 5mg
30333373 10 mg 10mg
30345898 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30345721 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCB2957555MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl (2-((((2,2-Dimethyl-1,3-dioxolan-4-yl)methoxy)(hydroxy)phosphoryl)oxy)ethyl)carbamate Sodium Salt
Analyte or Component Names Benzyl (2-((((2,2-Dimethyl-1,3-dioxolan-4-yl)methoxy)(hydroxy)phosphoryl)oxy)ethyl)carbamate Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C16 H23 N Na O8 P
InChI Formula InChI=1S/C16H24NO8P.Na/c1-16(2)22-11-14(25-16)12-24-26(19,20)23-9-8-17-15(18)21-10-13-6-4-3-5-7-13;/h3-7,14H,8-12H2,1-2H3,(H,17,18)(H,19,20);/q;+1/p-1
Solution Type Neat
SMILES CC1(C)OCC(COP(=O)(O[Na])OCCNC(=O)OCc2ccccc2)O1
IUPAC Name sodium;(2,2-dimethyl-1,3-dioxolan-4-yl)methyl 2-(phenylmethoxycarbonylamino)ethyl phosphate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 411.319
Quantity 5 mg
Formula Weight 411.106
Show More Show Less
Product Title
Select an issue

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.