Learn More
Benthiocarb, SPEX CertiPrep™
Product Code. 12939424 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12939424 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12939424 Supplier SPEX Certiprep Supplier No. S400
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
Benthiocarb28249-77-61000μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Packaging 2mL Amber Prescored Glass Ampule
CAS 28249-77-6, 67-56-1
CAS Min % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molecular Formula C12H16ClNOS
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key QHTQREMOGMZHJV-UHFFFAOYSA-N
SMILES CCN(CC)C(=O)SCC1=CC=C(C=C1)Cl
IUPAC Name S-[(4-chlorophenyl)methyl] N,N-diethylcarbamothioate
Solvent or Matrix Methanol-P&T
Molecular Weight (g/mol) 257.776
PubChem CID 34192
Quantity 1 mL
ChEBI CHEBI:35011
Chemical Name or Material Benthiocarb
Show More Show Less
Product intended for laboratory use only